Learn More
FLAD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555903
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RTDPYSCSLC PFSPTDPGWP AFMRINPLLD WTYRDIWDFL RQLFVPYCIL YDRGYTSLGS RENTVRNPAL KCLSPGGHPT YRPAYLLENE E Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
FLAD1 Polyclonal antibody specifically detects FLAD1 in Human samples. It is validated for Immunohistochemistry.Specifications
FLAD1 | |
Polyclonal | |
Unconjugated | |
FLAD1 | |
A930017E24Rik; fa92c06; FAD pyrophosphorylase; FAD synthase; FAD synthase region; FAD synthetase; FAD1; FAD1 flavin adenine dinucleotide synthetase homolog; Fad1, flavin adenine dinucleotide synthetase, homolog; FADS; FAD-synthetase; FLAD1; Flavin adenine dinucleotide synthase; flavin adenine dinucleotide synthetase; flavin adenine dinucleotide synthetase 1; FMN adenylyltransferase; hypothetical protein LOC506682; LSMFLAD; Molybdenum cofactor biosynthesis protein-like region; PP591; RFad1, flavin adenine dinucleotide synthetase, homolog (yeast); RP11-307C12.7; wu:fa92c06; zgc:91843 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFF5 | |
FLAD1 | |
Recombinant protein corresponding to Human FLAD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only