Learn More
Fisherbrand™ Vacuum Aspiration Advantage System Accessory, HandVac Duo Kit
For use with Vacuum Aspiration Advantage System
Brand: Fisherbrand™
Additional Details : Weight : 0.47000kg
Description
Kit Components:- Single channel plastic adapter for pipettor tip with ejector
- Silicone hose (inside diameter 6mm, wall thickness 3mm, length 2m)
- Rapid action coupling hose
- Connector and Y-connector
Specifications
Vacuum Aspiration Advantage System | |
HandVac Duo Kit |