Learn More
Fisherbrand™ Immersion Basket with Handle
Faciliates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™
Additional Details : Weight : 0.58950kg
 View more versions of this product
Product Code. 11756188
  • 65.30 EUR / Each
Estimated Shipment
18-03-2021
Add to basket
Description
Description
  • Designed for the Fisherbrand Dual Frequency Ultrasonic Cleaner models
  • Fine mesh bottom keeps small parts secure
  • Two handles for ease-of-use
  • Clean and simple design fits perfectly
  • Top-quality durable stainless steel
Specifications
Specifications
11.5 cm
Stainless Steel Basket
Stainless Steel
5.75 L Ultrasonic Cleaning Unit
7.5 cm
25.5 cm