Learn More
Fisherbrand™ Permanent Markers
Write on all surfaces
Brand: Fisherbrand™
Additional Details : Weight : 0.09900kg
Description
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
- Produce precise, ultrathin lines 0.3mm wide
- Waterproof and fadeproof, permanent ink dries almost instantly
- An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
- Unique stainless-steel long leg allows precise writing in a confined space