Learn More
Fisherbrand™ DNA Sequencing Systems: Glass Plates for Model FB-SEQ-3545
DNA Sequencing Systems Glass Plates for model FB-SEQ-3545
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11907515
  • 39.35 EUR / Each
Estimated Shipment: 25-07-2024
Description
For use with model FB-SEQ-3545
Specifications
Specifications
Sequencing gel casting systems
45 cm
Blank Glass
4.5 mm