Learn More
Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Wash Heads
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand:  Fisherbrand™
Additional Details : Weight : 0.01250kg
 View more versions of this product
Product Code. 11778006
  • 914.00 EUR / Each
Estimated Shipment: 05-06-2024
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Applications:
ELISA-based assays, cell washing
Specifications
Specifications
ELISA-based assays, cell washing
Wash Head
1 x 12
Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer