Learn More
Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand: Fisherbrand™
Additional Details : Weight : 0.28200kg
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Specifications
2L | |
Wellwash™ microplate strip washer | |
Cap with tubing and liquid level sensors |
67.62 oz., 2L | |
Waste Bottle |