Learn More
Traceable™ One-Shot™ pH Buffer Standards pH Certified Reference Material (CRM), Fisherbrand™
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisher Scientific Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
20.95 EUR
Product Code | Brand | Chemical Name or Material | pH | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Chemical Name or Material | pH | Quantity | Price | Quantity & Availability | |||
15568703
|
Fisherbrand
|
Buffer Standards One-Shot pH 10.012(CRM) | 10.012 | 6 x 100 mL |
20.95 EUR
100mL |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15558703
|
Fisherbrand
|
Buffer Standards One-Shot pH 7.000(CRM) | 7.00 | 6 x 100 mL |
20.95 EUR
100mL |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisher Scientific Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
- Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Supplied withindividual temperature chart, traceability information and Traceable™ Certificate