missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ 230V AC Adapter for Portable Meters

For accumet AP60 Series models AP65, AP64, AP63, AP62, AP61; with EUROPEAN PLUG

Brand:  Fisherbrand™

Additional Details : Weight : 0.10000kg

Product Code. 11510184

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

AC Adapter
European plug
Accumet™ AP60 AP65, AP64, AP63, AP62, AP61 portable meter
pH
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers