Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Product Code. 15863537 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Emission Filter
Emission Filter Long Pass
Excitation Filter
Excitation and Emission Filter
Fluorometric Emission Filter
Fluorometric Excitation Filter
Fluorometric Excitation and Emission Filter
Low Plate Adapter
Unit Size:
Each
Wavelength:
320/40 nm
335/13 nm
340 nm
350 nm
355 nm
360 nm
370/12 nm
380 nm
395 nm
400/10 nm
405 nm
410 nm
415 nm
420 nm
425 nm
430 nm
440 nm
444 nm
450/11 nm
460/24 nm
470 nm
475 nm
480/10 nm
485 nm
485/16 nm
490 nm
492/13 nm
510/10 nm
515 nm
517/15 nm
518/13 nm
520/10 nm
521 nm
525/30 nm
527 nm
530/10 nm
538/25 nm
540/10 nm
542 nm
550 nm
555/25 nm
560/10 nm
570 nm
575 nm
578 nm
584 nm
590/14 nm
597/8 nm
603/7 nm
604 nm
612 nm
620 nm
620/10 nm
621 nm
635 nm
640 nm
640/10 nm
646/10 nm
650 nm
670/10 nm
678 nm
680/20 nm
700 nm
790 nm
Fluoroskan™, Fluoroskan™FL, and Luminoskan™ microplate fluorometers and luminometers can be equipped with a wide range of accessories, additional excitation filters, and emission filters to expand the instrument's functionality and application flexibility.
For running fluorometric or luminometric assays, the Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers offer reliability and optical technology that has already been proven in thousands of laboratories all over the world. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility. Additionally, the Thermo Scientific™ SkanIt™ Software increases efficiency in your lab.
  • Filters play an essential role in fluorometric measurements. When ordering special filters, the better information you provide, the better the measurement results will be.
  • If you need advice on which filters to order, contact your local Thermo Fisher Scientific representative with the following details:
    • Name of the application
    • Excitation and emission spectra
    • Peak or center wavelengths of the excitation and emission spectra
  • Fluorometric filters can also be used for luminometry depending on the application. For luminometric applications requiring filters, contact technical support for instructions.
  • For further information, please read the following flyer Recommended filter pairs for various fluorescent dyes for Fluoroskan and Fluoroskan FL Microplate Fluorometers  
TRUSTED_SUSTAINABILITY
Specifications
Product Type Low Plate Adapter
For Use With (Equipment) Luminoskan™, Fluoroskan™ FL