Learn More
Gilson™ Filter for P5000 and P10mL Tips
Manufactured by Gilson™. Gilson™ Filter for P5000 and P10mL Tips are guaranteed to deliver value and assure reliability and precision.
Brand: Gilson™ F161241
38.86 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 0.04680kg
Description
- For use with P5000 and P10mL Pipetman™ Classic™
- Replacement filter
Specifications
Replacement filter | |
Filtered | |
Autoclavable | |
Yes | |
Polypropylene |
100 to 1000 μL | |
Sterile | |
Gilson™ Pipetman™ C1200, Gilson™ Pipetman™ U1000, Gilson™ Pipetman™ F250 to F1000, Gilson™ Pipetman™ P1000N, Gilson™ Pipetman™ P1000 | |
Diamond Sterlipack |