Learn More
BRAND™ Filter Funnel Supports
Polypropylene plates with aluminum support rod
Brand:  BRAND™ 74511
201.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10018020
  • 216.32 EUR / Each
Only null left
Estimated Shipment: 15-08-2025
Description
  • Manufactured from polypropylene plates with aluminum support rod
  • Adjustable funnel height
  • Support rod is 12.7 mm in diameter, 595 mm long
  • Provided individually
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
4 funnels
450 x 140 mm