Learn More
BRAND™ Filter Funnel Supports
Polypropylene plates with aluminum support rod
Brand:  BRAND™ 74506
Additional Details : Weight : 1.16000kg
 View more versions of this product
Product Code. 10212091
  • 164.00 EUR / Each
Estimated Shipment: 30-05-2024
Description
  • Manufactured from polypropylene plates with aluminum support rod
  • Adjustable funnel height
  • Support rod is 12.7 mm in diameter, 595 mm long
  • Provided individually
  • Specifications
    Specifications
    Polypropylene
    250 x 140 mm
    1
    2 funnels
    For 2 filter funnels