missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene™ UV Filter for 3 Position Filter Holder
Syngene™ UV Filter for 3 Position Filter Holder works in conjuction with the Syngene Gbox and Gbox Mini Systems.
Brand: Syngene™ FILTUV-GX
Additional Details : Weight : 0.20000kg
Description
- For use in GBOX and Gbox Mini systems 7 position filter wheel.
Specifications
Fixed | |
Gbox and Gbox Mini | |
572-625nm |