Learn More
Agilent BioTek Filter Cube
For use with Synergy LX
Brand:  Agilent BioTek 1505005
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15988550
  • 1025.00 EUR / 1 unit
Estimated Shipment: 14-06-2024
Specifications
Synergy LX
EX 485/20, EM 528/20, 510 nm mirror (Green)