missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Mouse anti-Human, Alexa Fluor 750, Clone: 427819, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB3715S-100UG
Additional Details : Weight : 0.00970kg
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry.Specifications
Fibroblast Activation Protein alpha/FAP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q12884 | |
FAP | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884 | |
RUO | |
2191 | |
Human | |
IgG1 |
Flow Cytometry | |
427819 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
Mouse | |
100 μg | |
Primary | |
Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only (RUO)