Learn More
Fibrinogen beta chain Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16747-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.Specifications
Fibrinogen beta chain | |
Polyclonal | |
Unconjugated | |
FGB | |
FGB, fibrinogen beta chain | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110135, 2244, 24366 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02675, P14480, Q8K0E8 | |
FGB | |
Fibrinogen beta chain Fusion Protein Ag10169 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |