Learn More
Eppendorf™ New Brunswick D81 Freezer Box Divider, 9 x 9 Matrix
For Freezer Models B50-SQ, B75-SQ & B95-SQ, 9 x 9 Matrix, Max Tube Diameter: 13.0mm. Allows freezer box to hold up to 81 vials or tubes.
Brand: Eppendorf™ D81
Additional Details : Weight : 4.54000kg