missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGFR3, His Tag Recombinant Protein
FGFR3 is a member of the FGFR family of receptor tyrosine kinases.
Brand: Invitrogen™ PV3145
Additional Details : Weight : 3.36000kg
Description
FGFR3 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.Specifications
NP_000133 | |
2261 | |
10 μg | |
FGF Receptor Kinase Family | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
FGFR3 | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
49.8 kDa | |
FGFR3, CEK2, JTK4 | |
aa 399-806 | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.2mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
fibroblast growth factor receptor 3 | |
Kinases & Inhibitors | |
Liquid | |
Oncology, Signal Transduction |