Learn More
Thermo Scientific™ BspPI (AlwI) (2 U/μL)
Cut at GGATC(4/5)^ sites with BspPI (AlwI) restriction enzyme, which performs best at 55°C in Tango buffer (Isoschizomers: AclWI, AlwI).
37.83 EUR - 158.00 EUR
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10384630
View Documents Promotion Details
Thermo Scientific™
ER1321
100 U
37.83 EUR
100 units
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
10569510
View Documents Promotion Details
Thermo Scientific™
ER1322
500 U
158.00 EUR
500 units
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities