Learn More
Cytiva Female Luer Lock Connector for ÄKTA Systems
Shop All Cytiva Products
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Cytiva Female Luer Lock Connector can be used with numerous ÄKTA liquid chromatography systems, including: ÄKTAexplorer™, ÄKTApurifier™, ÄKTAfplc™, or ÄKTAmicro™ systems.
Specifications
Specifications
| Type | Female Luer Lock Connector |
| Connections | Luer lok, female |
| Quantity | 1 Ea. |
| Color | Clear |
| For Use With (Equipment) | Autosampler A-900 and A-905, Used in Numerous ÄKTA Systems, Including: ÄKTAexplorer, ÄKTApurifier, Ä |