missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM105B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5143981
This item is not returnable.
View return policy
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
Otulin is a deubiquitinase that specifically removes linear ('Met-1'-linked) polyubiquitin chains to substrates and acts as a regulator of angiogenesis and innate immune response. Required during angiogenesis, craniofacial and neuronal development by regulating the canonical Wnt signaling together with the LUBAC complex. Acts as a negative regulator of NF-kappa-B by regulating the activity of the LUBAC complex. OTULIN function is mainly restricted to homeostasis of the LUBAC complex: acts by removing 'Met-1'-linked autoubiquitination of the LUBAC complex, thereby preventing inactivation of the LUBAC complex. Acts as a key negative regulator of inflammation by restricting spontaneous inflammation and maintaining immune homeostasis. In myeloid cell, required to prevent unwarranted secretion of cytokines leading to inflammation and autoimmunity by restricting linear polyubiquitin formation. Plays a role in innate immune response by restricting linear polyubiquitin formation on LUBAC complex in response to NOD2 stimulation, probably to limit NOD2-dependent proinflammatory signaling.Specifications
FAM105B | |
Polyclonal | |
Unconjugated | |
OTULIN | |
C79097; deubiquitinating enzyme otulin; FAM105B; family with sequence similarity 105, member B; GUM; gumby; m3Sapc; m7-1Sapc; OTU deubiquitinase with linear linkage specificity; OTU domain-containing deubiquitinase with linear linkage specificity; Otulin; protein FAM105B; ubiquitin thioesterase Gumby; Ubiquitin thioesterase otulin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100362554, 432940, 90268 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4MG trehalose and 0.01MG sodium azide | |
Q3UCV8, Q96BN8 | |
OTULIN | |
E.coli-derived human OTULIN recombinant protein (Position: A49-L352). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |