missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4B10, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | FADS2 |
---|---|
Clone | 1G4B10 |
Concentration | 1000 μg/mL |
Applications | Immunohistochemistry, Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17721139
|
Proteintech
68026-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17711139
|
Proteintech
68026-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FADS2 Monoclonal antibody specifically detects FADS2 in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISASpecifications
FADS2 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat | |
9415 | |
Primary | |
Store at -20°C |
1G4B10 | |
Immunohistochemistry, Western Blot, ELISA | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
IgG2b | |
Protein A purification | |
Antibody |