Learn More
Motic™ Widefield Eyepiece WF20X/11mm
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF20X/11mm provides an improved field of view and greater eye relief.
Brand: Motic™ 1101001400572
Additional Details : Weight : 0.01000kg
Description
- Widefield style provides improved field of view and greater eye relief.
- Magnification: 20X.
- Field of view: 11mm.
Specifications
Widefield | |
FOV: 11mm | |
DS-2, DM-111, DM-1802, DMW-B1, F-11, 18 Series, 28 Series, PM Series, B1 Series, B3 Series Microscopes | |
20X |