Learn More
Motic™ Widefield Eyepiece WF20X/11mm
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece WF20X/11mm provides an improved field of view and greater eye relief.
Brand:  Motic™ 1101001400572
Additional Details : Weight : 0.01000kg
Product Code. 15445082
  • 53.00 EUR / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Widefield style provides improved field of view and greater eye relief.
  • Magnification: 20X.
  • Field of view: 11mm.
Specifications
Specifications
Widefield
FOV: 11mm
DS-2, DM-111, DM-1802, DMW-B1, F-11, 18 Series, 28 Series, PM Series, B1 Series, B3 Series Microscopes
20X