Learn More
JULABO™ Extendable Bridge
Extendable from 330 to 680 mm
Brand:  JULABO™ 9970201
Additional Details : Weight : 1.30000kg
Product Code. 15480612
  • 451.00 EUR / Each
Estimated Shipment: 02-07-2024
Description
This accessory is suitable for:
CORIO™ CD, CORIO™ CP, DYNEO™ DD
Specifications
Specifications
CORIO™ CD, CORIO™ CP, DYNEO™ DD
Extendable bridge
330 mm to 680 mm