Learn More
Gibco™ Expi293F™ Inducible Cells
Expi293F Inducible cell line is derived from engineered Expi293F cells that stably express the tetracycline repressor (TetR) protein, thus enabling tetracycline-regulated protein expression
Brand: Gibco™ A39241
2313.25 EUR valid until 2024-12-31
Use promo code "21941" to get your promotional price.
Additional Details : Weight : 0.00300kg
Description
Cell line established by the stable integration of the regulatory plasmid pcDNA6/TR, which encodes the TetR gene under the control of the human CMV promoter. It is a powerful tool for tetracycline-inducible high-yield expression of recombinant proteins.
Features:
- Suspension, high-density culture in Expi293 Expression Medium
- Tetracycline-inducible protein expression with extremely low basal expression levels
- Equivalent protein yields to parental Expi293F cells—up to 1 g/L
- Significantly higher protein yields than other HEK293 Tet-inducible cell line alternatives
For inducible expression of a protein of interest, pcDNA5/TO mammalian expression vector or an equivalent inducible vector is required for tetracycline-regulated gene expression.
Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium
Specifications
1 vial Expi293F Inducible Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen |
|
Expi293™ | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.