Learn More
EVOS™ EVOS™ 20X Objective, achromat, LWD, phase-contrast
Components manufactured to ensure publication-quality images
Brand: EVOS™ AMEP4634
Additional Details : Weight : 0.01000kg
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
EVOS LWD Achromatic Phase Contrast Objective | |
EVOS™ XL Imaging System, EVOS™ XL Core Imaging System | |
Plan Achromat | |
0.40 |
EVOS | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
20X | |
1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.