Learn More
Ethyl 4-chlorobutyrate, 97%, Thermo Scientific Chemicals
Catalog Number p-3757826
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3153-36-4
C6H11ClO2
150.6
MFCD00001004
OPXNFHAILOHHFO-UHFFFAOYSA-N
ethyl 4-chlorobutyrate, butanoic acid, 4-chloro-, ethyl ester, ethyl gamma-chlorobutyrate, ethyl-4-chlorobutyrate, 4-chloro-butyric acid ethyl ester, butyric acid, 4-chloro-, ethyl ester, ethyl .gamma.-chlorobutyrate, 4-chlorobutyric acid ethyl ester, ethyl 4chlorobutyrate, acmc-1cqjw
18474
ethyl 4-chlorobutanoate
CCOC(=O)CCCCl
3153-36-4
C6H11ClO2
150.6
MFCD00001004
OPXNFHAILOHHFO-UHFFFAOYSA-N
ethyl 4-chlorobutyrate, butanoic acid, 4-chloro-, ethyl ester, ethyl gamma-chlorobutyrate, ethyl-4-chlorobutyrate, 4-chloro-butyric acid ethyl ester, butyric acid, 4-chloro-, ethyl ester, ethyl .gamma.-chlorobutyrate, 4-chlorobutyric acid ethyl ester, ethyl 4chlorobutyrate, acmc-1cqjw
18474
ethyl 4-chlorobutanoate
CCOC(=O)CCCCl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Brown to Yellow
Density 1.0700g/mL
Boiling Point 186.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3CO2C2H5
Refractive Index 1.4296 to 1.4316
Quantity 5 g
Beilstein 02, 278
Specific Gravity 1.07
Solubility Information Solubility in water: immiscible
Formula Weight 150.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 4-chlorobutyrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 4-chlorobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.