missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Boiling Point | 207°C to 208°C |
Refractive Index | 1.5225 |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 156.2 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl thiophene-3-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'