missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals

Catalog Number 11450637
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
5751-80-4
C7H8O2S
156.199
MFCD02179226
OYSLMAQEMAJMCL-UHFFFAOYSA-N
ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n
79824
ethyl thiophene-3-carboxylate
CCOC(=O)C1=CSC=C1
This item is not returnable. View return policy
5751-80-4
C7H8O2S
156.199
MFCD02179226
OYSLMAQEMAJMCL-UHFFFAOYSA-N
ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n
79824
ethyl thiophene-3-carboxylate
CCOC(=O)C1=CSC=C1

Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 207°C to 208°C
Refractive Index 1.5225
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 156.2
Percent Purity 98%
Chemical Name or Material Ethyl thiophene-3-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'