missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(chlorosulfonyl)-3-furoate, 95%, Thermo Scientific™

Catalog Number p-3629716
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
256373-91-8
C7H7ClO5S
238.638
MFCD11109333
LEKOVVUPHAKDKN-UHFFFAOYSA-N
ethyl 5-chlorosulfonyl furan-3-carboxylate, ethyl 5-chlorosulfonyl-3-furoate, 3-furancarboxylic acid,5-chlorosulfonyl-, ethyl ester, 5-chlorosulfonyl-furan-3-carboxylic acid ethyl ester, 2-chlorosulfonyl furan-4-carboxylic acid ethyl ester, 3-furancarboxylic acid, 5-chlorosulfonyl-, ethyl ester
10130959
ethyl 5-chlorosulfonylfuran-3-carboxylate
CCOC(=O)C1=COC(=C1)S(=O)(=O)Cl
This item is not returnable. View return policy
256373-91-8
C7H7ClO5S
238.638
MFCD11109333
LEKOVVUPHAKDKN-UHFFFAOYSA-N
ethyl 5-chlorosulfonyl furan-3-carboxylate, ethyl 5-chlorosulfonyl-3-furoate, 3-furancarboxylic acid,5-chlorosulfonyl-, ethyl ester, 5-chlorosulfonyl-furan-3-carboxylic acid ethyl ester, 2-chlorosulfonyl furan-4-carboxylic acid ethyl ester, 3-furancarboxylic acid, 5-chlorosulfonyl-, ethyl ester
10130959
ethyl 5-chlorosulfonylfuran-3-carboxylate
CCOC(=O)C1=COC(=C1)S(=O)(=O)Cl
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.65
Percent Purity 95%
Chemical Name or Material Ethyl 5-(chlorosulfonyl)-3-furoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'