missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-furoate, 99%, Thermo Scientific Chemicals

Catalog Number p-7021536
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
614-99-3
C7H8O3
140.138
MFCD00003237
NHXSTXWKZVAVOQ-UHFFFAOYSA-N
11980
ethyl furan-2-carboxylate
CCOC(=O)C1=CC=CO1
This item is not returnable. View return policy
614-99-3
C7H8O3
140.138
MFCD00003237
NHXSTXWKZVAVOQ-UHFFFAOYSA-N
11980
ethyl furan-2-carboxylate
CCOC(=O)C1=CC=CO1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 31°C to 36°C
Density 1.12
Boiling Point 195°C to 196°C
Flash Point 70°C (158°F)
Quantity 25 g
Beilstein 2653
Merck Index 14,4307
Synonym ethyl 2-furoate, ethyl furoate, ethyl 2-furancarboxylate, 2-furancarboxylic acid, ethyl ester, ethyl pyromucate, 2-carboethoxyfuran, furan-2-carboxylic acid ethyl ester, ethyl-2-furoate, 2-furoic acid, ethyl ester, ethyl2-furoate
Formula Weight 140.14
Percent Purity 99%
Chemical Name or Material Ethyl 2-furoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'