Learn More
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
Solubility
Sparingly soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
Solubility
Sparingly soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Quantity | 250 mg |
Beilstein | 4426004 |
Solubility Information | Sparingly soluble in water. |
Formula Weight | 274.13 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate |