missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7027172
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
594-44-5
C2H5ClO2S
128.57
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(Cl)(=O)=O
This item is not returnable. View return policy
594-44-5
C2H5ClO2S
128.57
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride, ethylsulfochloride, 1-ethanesulfonyl chloride, ethanesulphonyl chloride, ethanesulfonylchloride, unii-b56zoc898a, ethylsulfonylchloride, ethansulfonyl chloride, ethansulfonylchlorid, ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70°C
Density 1.35
Boiling Point 176°C to 178°C
Flash Point 83°C (181°F)
Linear Formula CH3CH2SO2Cl
Refractive Index 1.454
Quantity 25 g
UN Number UN2927
Beilstein 773865
Sensitivity Moisture sensitive
Formula Weight 128.57
Percent Purity ≥98%
Chemical Name or Material Ethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'