missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B, ∽93% by uv-vis, MP Biomedicals™

Catalog Number p-4405532
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
568-63-8
C40H14I8Na2O10
1715.757
MFCD00144257
DKRYETBOLGDIOK-UHFFFAOYSA-L
Acid Red 51, C.I. 45430
131676154
disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
568-63-8
C40H14I8Na2O10
1715.757
MFCD00144257
DKRYETBOLGDIOK-UHFFFAOYSA-L
Acid Red 51, C.I. 45430
131676154
disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]
Assay Percent Range ∽ 93%
Formula Weight 879.9
Color Maroon
Quantity 10 g
Physical Form Powder
Chemical Name or Material Erythrosin B Sodium Salt
compliance-icons
Product Identifier
  • Erythrosin B Sodium Salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'