Learn More
Eppendorf™ 5810 Series 4 x 750mL Swing-bucket Rotor
Accepts a variety of bucket types
1580.00 EUR - 3385.00 EUR
Specifications
Capacity | 4 x 750 mL |
---|---|
Max. RCF | 3,214 x g |
Max. Speed | 3,900 rpm |
For Use With (Equipment) | 5810/5810R Centrifuge |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12806233
|
Eppendorf™
5820740000 |
With Round Buckets |
3385.00 EUR
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15214536
|
Eppendorf™
5820755008 |
Without Buckets |
1580.00 EUR
Each |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12876233
|
Eppendorf™
5820754001 |
With Buckets for Deepwell Plates |
3385.00 EUR
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Capacity: 3L (4 x 750mL)
- Max. Speed/RCF: 3,900rpm/3,214 x g
Specifications
4 x 750 mL | |
3,214 x g | |
3,900 rpm | |
5810/5810R Centrifuge |