Learn More
Motic™ Epi-Fluorescence Attachment with Filter Cassette, BA410E, BA310E, AE31E
Easily integrated into the BA410 arm. Motic™ Epi-Fluorescence Attachment with Filter Cassette, BA410E, BA310E, AE31E accommodates four filter cassettes for excitation observation.
Brand:  Motic™ 1101000202161
Additional Details : Weight : 0.01000kg
Product Code. 15445302
  • 1620.00 EUR / Each
Estimated Shipment: 26-07-2024
Description
  • Four cassette capacity.
  • Slider format for cassette change.
Specifications
Specifications
Epi-fluorescence Attachment
BA410E, BA310E, AE31E Microscopes
Filter cassette