missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eosin Y Disodium Salt, MP Biomedicals
Click to view available options
Quantity:
25 g
100 g
250 g
Specifications
Formula Weight | 691.86 |
Color | Red |
Quantity | 25 g |
Physical Form | Powder |
Chemical Name or Material | Eosin Y Disodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'