Learn More
DWK Life Sciences DURAN™ Blank for Engler Distilling Flask, with beaded rim
For further processing.
Brand:  DWK Life Sciences 216512402
Additional Details : Weight : 0.68000kg
 View more versions of this product
Product Code. 15281779
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Blanks for DURAN™ Engler Distilling Flasks are intended for further processing into DURAN™ Engler flask according to ASTM D86 and DIN EN ISO 3405. Engler flasks are ideal as heating glasses and for distillations due to their uniform wall thickness.
  • With beaded rim
  • For further processing to DURAN™ Engler flask according to ASTM D86 and DIN EN ISO 3406
  • Due to uniform wall thickness ideal as cooking glass and suitable for distillations
  • Glass articles for further processing
Specifications
Specifications
66 mm
Borosilicate Glass (3.3)
10/Pk.
100 mL
For further processing.; Heating
Yes
Product Suggestions
Product Suggestions
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote