Learn More
Merck™ Syringe
Dispense liquids into small containers and pressure-filter small volumes through filter holders or filter units with female Luer slip inlets.
Brand:  Merck™ XX1102012
Additional Details : Weight : 0.80000kg
Product Code. 11973906
  • 97.15 EUR / Pack of 12
Estimated Shipment
02-08-2019
Add to basket
Description
Description
  • Polypropylene syringe
  • Male Luer slip outlet
  • 20mL capacity
Order Info
12 per pack
Specifications
Specifications
20mL
Polypropylene
Syringe
For dispensing and filtering small liquid volumes
Luer-slip