Learn More
Merck Replacement Solutions for Montage™ BAC96 Miniprep Kit and Montage Plasmid Miniprep96 Kit
Suitable for cell resuspension for plasmid & BAC minipreps
Brand: Merck LSKCRS500
155.66 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.36800kg
Description
The Montage Plasmid MiniprepHTS 96 Kit is a fast, easy-to-use kit for high-purity plasmid or bacterial artificial chromosome (BAC) miniprepsSpecifications
Plasmid Miniprep, BAC Prep | |
Cell Resuspension Solution | |
500 mL |