Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Product Code. 10659303 Shop All Merck Products
Click to view available options
Color:
Clear
White
Material:
Acrylic
Polystyrene
Sterility:
Non-sterile
Sterile
Membrane:
Hydrophobic PVDF
Hydrophobic PVDF with Underdrain
Mixed Cellulose Esters
Unit Size:
Pack of 10
Pack of 50
Product Code. 10659303
Brand: Merck MAIPSWU10
Only null left
Add to basket
Add to basket
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Yes
Membrane Hydrophobic PVDF
Array 8 x 12
Pore Size 0.45 μm
Product Line MultiScreen
Color White
Product Type MultiScreen IP
Material Acrylic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) ELISPOT Assays
Quantity 10/Pk.