missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Microcon™ Centrifugal Filters for Protein and DNA Concentration: Regenerated Cellulose Membrane
ideal for DNA concentration and desalting applications.
Brand: Merck Millipore MRCF0R100
Additional Details : Weight : 0.28350kg
Description
ideal for DNA concentration and desalting applications.
Non sterileFilter Type: Ultrafiltration
Application: Nucleic acid purification
Length: 4.50cm
Diamter: 12.3mm
Filtration area: 0.32 sq. cm
Volume: 0.5mL
Specifications
Non-sterile | |
0.5 mL | |
Blue | |
4.5cm | |
0.32cm2 |
Centrifugal Filter | |
Regenerated Cellulose | |
12.3mm | |
100 kDa |