Learn More
Merck Accessories for Dispensing Pressure Vessels
Accessory for use with Dispensing Pressure Vessels
146.00 EUR - 1830.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
For Use With (Equipment) | Dispensing Pressure Vessels |
Sterility | Non-sterile |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10355091
|
Merck
XX6700035 |
Tubing |
146.00 EUR
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10717871
|
Merck
XX6700059 |
O-Ring |
185.00 EUR
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
10209101
|
Merck
XX67000PK |
Pressure Vessel Accessory Kit |
1830.00 EUR
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10518411
|
Merck
XX6700L55 |
Bubble Point Test Apparatus |
1620.00 EUR
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Specifications
Not Autoclavable | |
Dispensing Pressure Vessels | |
Non-sterile |