Learn More
Motic™ Phase Stop 0.5 Ph1 (10X, 20X, 40X)
Use phase contrast to improve visualization of transparent specimens. Motic™ Phase Stop 0.5 Ph1 (10X, 20X, 40X) is an annular light stop that fits in your condenser.
Brand:  Motic™ 1101000202421
Additional Details : Weight : 0.01000kg
Product Code. 15465362
  • 37.00 EUR / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
  • Must be matched and aligned to the objective phase plate.
Specifications
Specifications
Phase Stop Annulus
0.5 Ph1 (10X, 20X, 40X)
AE31E Microscope, 10X, 20X, 40X phase objectives