Learn More
LTE Scientific™ Electric Hoist
An accessory item for use with all top-loading Touchclave autoclave models.
Brand: LTE Scientific™ TC/HST/01
Additional Details : Weight : 1.00000kg
Description
- Aids in the sterilization process
- Convenient method of lowering heavy loads into top-loading autoclaves
Specifications
Top-loading Touchclave autoclave models | |
Electric Hoist |