Learn More
Invitrogen™ EIF2AK2 (PKR) Recombinant Human Protein
EIF2AK2 (PKR) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha.
Brand: Invitrogen™ PV4822
Additional Details : Weight : 0.01000kg
Description
EIF2AK2 (PKR) phosphorylates Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses. EIF2AK2 plays a key role in antiviral defense mediated through its direct activation by double-stranded RNA produced by viral infection.
Specifications
NP_002750.1 | |
50mM Tris, pH 7.5, 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT | |
1mg | |
RUO - research use only | |
EIF2AK2 (PKR) | |
aa 252-551 | |
Recombinant | |
Baculovirus |
Nuclear Receptor Assay | |
62.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
PEK Kinase Family | |
Human | |
GST Tag | |
Frozen |