missing translation for 'onlineSavingsMsg'
Learn More

EGFR Recombinant Human Protein, His Tag, Active

EGFR Recombinant Human Protein (His Tag, Active)

Brand:  Invitrogen 10001H08H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15284475

  • 165.00 EUR / 25µg


Estimated Shipment date
from Supplier 29-10-2018
Add to basket

Description

Description

Epidermal growth factor receptor (EGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant protein

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) fused to a polyhistidine tag at the C-terminus.

Activity: Measure by its ability to bind with human EGF in a functional ELISA.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

Specifications

Specifications

Protein assays and analysis
Lyophilized
EGFR Recombinant Human Protein
5 x 5μg
For research use only. Not for use in diagnostic procedures
Growth factor receptors, receptor tyrosine kinases
EGFR, ErbB, HER1, mENA, ErbB1, PIG61
EGFR
Growth factors and receptors, kinases and inhibitors
EGF Receptor (ErbB) kinase family
Human
Lyophilized
69.8kDa
>95% by SDS-PAGE
–20°C
1 Each
<1 EU/μg
EGFR Recombinant Human Protein
Assay standard
Mammalian
EGFR Recombinant Human Protein
His Tag
Videos