missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ ECO AluRack for Upright Freezers, 32 mm Box Height

ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11138

Additional Details : Weight : 1.90000kg

 View more versions of this product

Product Code. 12432490

  • 97.70 EUR / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Without white cardboard boxes
  • Holds is expressed by Height x Dept
Specifications

Specifications

Seawater resistant
Boxes (36)
Upright Freezers
32 mm (Box)
9 x 4
Aluminum
564 x 139 x 324 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers