Learn More
Tenak™ ECO AluRack for Chest Freezers, 75 mm Box Height
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
58.50 EUR - 71.55 EUR
Specifications
Description | Seawater resistant |
---|---|
Material | Aluminum |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 75 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408140
|
Tenak™
TE11538 |
Boxes (6) |
65.90 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12428130
|
Tenak™
TE11530 |
Boxes (2) |
58.50 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
12428140
|
Tenak™
TE11540 |
Boxes (7) |
69.30 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12448130
|
Tenak™
TE11532 |
Boxes (3) |
58.50 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12448140
|
Tenak™
TE11542 |
Boxes (8) |
71.55 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12468130
|
Tenak™
TE11534 |
Boxes (4) |
61.35 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12488130
|
Tenak™
TE11536 |
Boxes (5) |
63.60 EUR
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without standard white cardboard boxes
Specifications
Seawater resistant | |
Aluminum | |
Chest Frseezers | |
75 mm (Box) |